Skip to Content
Merck
All Photos(1)

Key Documents

Z743616

Sigma-Aldrich

Color coded Tough-Tags, tube and spot labels

white to yellow, L × W × diam. 1.25 in. × 0.5625 in. × 0.44 in., roll of 500

Synonym(s):

solvent resistant labels, temperature resistant labels

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
55121611
NACRES:
NB.35

packaging

roll of 500

Quality Level

manufacturer/tradename

COLY-1000

L × W × diam.

1.25 in. × 0.5625 in. × 0.44 in.

color

white to yellow

Looking for similar products? Visit Product Comparison Guide

General description

COLOR-CODED TOUGH-TAGS
  • Easily differentiate samples and identify what you need
  • Translucent writing area keeps contents visible
  • Repositionable for up to thirty minutes
  • Adhesive strengthens over time

Legal Information

Tough-Tags is a trademark of Diversified Biotech

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service