Skip to Content
Merck
All Photos(1)

Key Documents

VIV102015SB3

SynVivo SynBBB 3D blood brain barrier model chip, Transepithelial/endothelial Electrical Resistance (TEER) configuration

new
Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
60104506
NACRES:
NA.84

packaging

case of 3 ea

manufacturer/tradename

SynVivo 102015-SB3

storage temp.

room temp

General description

SynBBB Blood Brain Barrier Model Chip - TEER configuration - Barrier type: Slits, Slit width: 3 μm, Distance between slits: 50 μm, Travel/barrier width: 50 μm, Outer Channel width: 200 μm, Device depth: 100 μm, Impedence measurement. This TEER enabled chip is used to create the SynBBB model in monoculture, co or tri-culture configurations.

SynVivo’s SynBBB 3D blood brain barrier model recreates the in vivo microenvironment by emulating a histological slice of brain tissue cells in communication with endothelial cells across the blood brain barrier (BBB). Shear-induced endothelial cell tight junctions, which cannot be achieved in the Transwell model, are easily achieved in the SynBBB model using physiological fluid flow. Formation of tight changes can be measured using biochemical or electrical analysis (assessing changes in electrical resistance) with the SynVivo Cell Impedance Analyzer. Interactions between brain tissue cells and endothelial cells are readily visualized in the SynBBB assay. Transwell models do not allow real-time visualization of these cellular interactions, which are critical for understanding of the BBB microenvironment.

SynBBB is the only in vitro BBB model that allows:

  • Accurate in vivo hemodynamic shear stres
  • Real-time visualization of cellular and barrier functionality
  • Significant reduction in cost and time
  • Robust and easy to use protocols
The SynBBB system is a highly versatile platform for investigation of:

  • Tight junction proteins: Determine the levels of tight junction proteins namely zonula occludens, claudins and occludins which regulate the BBB.
  • Transporter proteins: Analyze functionality of transporter proteins (e.g. Pgp) in normal and dysfunctional BBB.
  • Drug permeability: Evaluate real-time permeability of therapeutics and small molecules across the endothelium of the BBB.
  • Inflammation: Understand the underlying mechanisms of inflammatory responses on the regulation of the BBB.
  • Cell migration: Visualize and quantify in real-time migration of immune cells across the BBB.
  • Omic changes: Perform genomic, proteomic and metabolic analysis on normal and dysfunctional BBB.
  • Neurotoxicity: Analyze toxicity effects of chemical, biological and physical agents on the cells of the BBB.
  • Neuro-oncology: Investigate effects of the tumor cells on the BBB.

Linkage

LInk to Publications application text

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service