VIV102012STU3
SynVivo SynTumour 3D cancer model chip, IMN2 radial configuration, 8 um pillar
All Photos(1)
About This Item
UNSPSC Code:
60104506
NACRES:
NA.84
packaging
case of 3 ea
manufacturer/tradename
SynVivo 102012-STU3
storage temp.
room temp
General description
SynTumor cancer model chip-Barrier type: Pillars, Distance between pillars: 8 μm, Barrier height: 8 μm, Travel/barrier width: 50 μm, Outer Channel width: 200 μm, Device depth: 100 um. SynTumor is a 3D tissue model for real-time visualization and quantitative assessment of cell-cell and cell-drug interactions in a physiologically and morphologically realistic tumor microenvironment.
Linkage
LInk to Publications application text
Choose from one of the most recent versions:
Certificates of Analysis (COA)