VIV102008SR3
SynVivo SynRAM 3D Inflammation model chip, IMN2 radial configuration
All Photos(3)
About This Item
UNSPSC Code:
60104506
NACRES:
NA.84
packaging
pkg of 3 ea
manufacturer/tradename
SynVivo 102008-SR3
storage temp.
room temp
General description
SynRAM Inflammation model chip - Barrier type: Pillars, Distance between pillars: 8 μm, Barrier height: 3 μm, Travel/barrier width: 50 μm, Outer Channel width: 200 μm, Device depth: 100 μm. This chip is used to create the SynRAM inflammation model in monoculture or co-culture configurations
Linkage
LInk to Publications application text
Choose from one of the most recent versions:
Certificates of Analysis (COA)