Skip to Content
Merck
All Photos(1)

Key Documents

SRP0682

Sigma-Aldrich

IGF1RTIDE

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

Assay

≥95% (HPLC)

form

lyophilized powder

mol wt

1595.83

shipped in

ambient

storage temp.

−20°C

Biochem/physiol Actions

The 14 amino acids of IGF1Rtide peptide sequence (KKKSPGEYVNIEFG) is derived from human IRS-1 protein residues 891-902.

Sequence

KKKSPGEYVNIEFG

Reconstitution

Reconstitute in 25 mM Tris-HCl, pH 7.5 solution to a final concentration of 1mg/mL.

Storage and Stability

Store diluted product in aliquots at -20°C. Avoid freeze/thaw cycles.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service