Skip to Content
Merck
All Photos(1)

Key Documents

SRP0369

Sigma-Aldrich

MNK1 active human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE)

Synonym(s):

MAP kinase interacting serine/threonine kinase 1

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

aqueous solution

mol wt

74 kDa

packaging

pkg of 10 μg

manufacturer/tradename

Croda International PLC

technique(s)

activity assay: suitable
inhibition assay: suitable

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MKNK1(8569)

General description

Human MNK (T385D) (GenBank Accession No. NM_003684), amino acids 2-End (amino acid deletion 165-205) with N-terminal GST-tag, MW=74 kDa, expressed in a Baculovirus infected Sf9 cell expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Physical form

Solution in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% TWEEN®-20, 30% glycerol, and 3 mM DTT.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Theodore Vougiouklakis et al.
Oncotarget, 6(41), 43162-43171 (2015-11-21)
The oncogenic protein ERK, a member of the extracellular signal-regulated kinase (ERK) cascade, is a well characterized signaling molecule involved in tumorigenesis. The ERK signaling pathway is activated in a large proportion of cancers and plays a critical role in
Bong Seok Kang et al.
Journal of cellular biochemistry, 118(8), 2357-2370 (2017-01-21)
ERK1 and ERK2 share a great deal of homology and have been presumed to have similar functions. Available antibodies recognize both isoforms making the elucidation of functional differences challenging. Mitogen-activated protein (MAP) kinase networks are commonly depicted in the literature
Seung-Hee Chang et al.
American journal of respiratory cell and molecular biology, 35(5), 528-539 (2006-06-10)
Inorganic phosphate (Pi) plays a critical role in diverse cellular functions. Among three classes of sodium/phosphate co-transporters (NPTs), two types have been identified in mammalian lung. The potential importance of Pi as a novel signaling molecule and pulmonary expression of
S G Gregory et al.
Nature, 441(7091), 315-321 (2006-05-20)
The reference sequence for each human chromosome provides the framework for understanding genome function, variation and evolution. Here we report the finished sequence and biological annotation of human chromosome 1. Chromosome 1 is gene-dense, with 3,141 genes and 991 pseudogenes
Xuesong Song et al.
Oncology reports, 34(3), 1573-1580 (2015-07-03)
MicroRNAs (miRNAs) are small non-coding RNAs that play critical roles in cervical carcinogenesis and progression. microRNA-133a (miR-133a) has been reported to play a tumor-suppressor role in a range of cancers. However, the role and underlying molecular mechanism of miR-133a in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service