Skip to Content
Merck
All Photos(1)

Key Documents

SML4132

Tafamidis meglumine

new

≥98% (HPLC)

Synonym(s):

2-(3,5-Dichlorophenyl)-1,3-benzoxazole-6-carboxylic acid mono (1-deoxy-1-methylamino-D-glucitol), Fx 1006A, Fx-1006A, Fx1006A, PF 06291826, PF-06291826, PF06291826

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H24Cl2N2O8
CAS Number:
Molecular Weight:
503.33
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to pink

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

Biochem/physiol Actions

Transthyretin (TTR) tetramer stabilizer.
Tafamidis is a transthyretin (TTR) tetramer stabilizer that binds selectively and with negative cooperativity to the two normally unoccupied thyroxine-binding sites of the tetramer (Kd ~2 nM and ~200 nM), kinetically stabilizing the wild-type as well as the variant TTR complexes (V30M, F64S, Y69H, I84S, V122I, and L111M) and preventing their dissociation into fibril-forming monomers.

Caution

Hygroscopic

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service