Skip to Content
Merck
All Photos(1)

Key Documents

SML4124

Ginsenoside Rb2

new

≥98% (HPLC)

Synonym(s):

(3β,12β)-20-[(6-O-α-L-Arabinopyranosyl-β-D-glucopyranosyl)oxy]-12-hydroxydammar-24-en-3-yl 2-O-β-D-glucopyranosyl-β-D-glucopyranoside, G-Rb2, Ginsenoside C, NSC 308878

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H90O22
CAS Number:
Molecular Weight:
1079.27
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Ginsenoside Rb2 exhibits diverse biological activities through interaction with various cellular receptors

Ginsenoside Rb2, a steroid glycoside found in Panax ginseng, demonstrates diverse biological activities. Exhibits adaptogenic properties, functions as an antioxidant, possesses anti-inflammatory, antidiabetic, antitumor, anti-osteoporosis and antimetastatic characteristics, provides neuroprotective effects, and acts as an immune modulator. Ginsenoside-Rb2 exerts modulation on various signaling pathways, including PI3K/Akt, MAPK, Nrf2/HO-1, Wnt/β-catenin, and AMPK pathways. Additionally, G-Rb2 activates SIRT-1 while antagonizing P2Y12R.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service