Skip to Content
Merck
All Photos(1)

Key Documents

SML4121

Erythrodiol

new

≥98% (HPLC)

Synonym(s):

3β-Erythrodiol, Oleanolic alcohol, (3β)-Olean-12-ene-3,28-diol, Olean-12-ene-3β,28-diol, (+)-Erythrodiol, 3β,28-Dihydroxyolean-12-ene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H50O2
CAS Number:
Molecular Weight:
442.72
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

Biochem/physiol Actions

Natural pentacyclic triterpenoid that exhibits anti-inflammatory, antioxidant, and antitumor properties, and enhances cholesterol efflux.

Erythrodiol, also known as Oleanolic alcohol, is a natural triterpene found in olive oil and other plants, known for its multi-faceted pharmacological actions. It displays anti-inflammatory effects by inhibiting pro-inflammatory cytokines and modulating enzymes involved in inflammation. As an antioxidant, it scavenges free radicals, mitigating oxidative stress. In addition, erythrodiol exhibits antitumor properties through antiproliferative activities like inducing cancer cell apoptosis and cell cycle arrest. Erythrodiol also enhances cholesterol efflux in macrophages by increasing the half-life of the ATP-binding cassette transporter A1 (ABCA1) protein, primarily by preventing its degradation.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service