Skip to Content
Merck
All Photos(1)

Key Documents

SML4015

Panobinostat lactate

new

≥98% (HPLC)

Synonym(s):

(2E)-N-Hydroxy-3-[4-[[[2-(2-methyl-1H-indol-3-yl)ethyl]amino]methyl]phenyl]-2-propenamide, LBH 589 lactate, LBH-589 lactate, LBH589 lactate, NVP-LBH589 lactate, N-Hydroxy-3-[4-[[[2-(2-methyl-1H-indol-3-yl)-ethyl]-amino]methyl]phenyl]-2E-2-propenamide, lactate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23N3O2·C3H6O3
CAS Number:
Molecular Weight:
439.50
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Orally active, potent, cinnamoyl hydroxamate-type pan-histone deacetylase inhibitor with anti-cancer efficacy in vitro and in vivo.

Panobinostat (LBH589) is an orally active, potent, cinnamoyl hydroxamate-type pan-histone deacetylase inhibitor (class I HDAC1/2/3/8 IC50 = 2.5/13.2/2.1/277 nM; IIA HDAC4/5/7/9 IC50 = 203/7.8/531/5.7 nM; IIB HDAC6/10 IC50 = 10.5/2.3 nM, IV HDAC11 IC50 = 2.7 nM) with anti-cancer efficacy in vitro and in vivo.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service