SML3888
Dynapyrazole Compound 20
≥98% (HPLC)
Synonym(s):
6-(3-bromophenoxy)-2- [1-(4-chlorophenyl) cyclopropyl]-7-oxo-4H-pyrazolo[1,5-a]pyrimidine-3-carbonitrile, Compound 20
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C22H14BrClN4O2
Molecular Weight:
481.73
UNSPSC Code:
12352200
NACRES:
NA.77
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
-10 to -25°C
Biochem/physiol Actions
Dynapyrazole Compound 20 is a potent and selective inhibitor of cytoplasmic S. cerevisiae dynein, an AAA (ATPase associated with diverse cellular activities), which engages the regulatory ATPase sites in the AAA3 and AAA4 domains disrupting allosteric communication across the AAA ring. Ddynapyrazole Compound 20 inhibits dynein’s basal ATPase activity. Also, it inhibits dynein motility by reducing the protein’s ability to bind microtubules.
Potent and selective inhibitor of cytoplasmic S. cerevisiae dynein
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service