Skip to Content
Merck
All Photos(1)

Key Documents

SML2994

Sigma-Aldrich

HQ461 Hydrobromide

≥98% (HPLC)

Synonym(s):

2-(2-((6-Methylpyridin-2-yl)amino)thiazol-4-yl)-N-(5-methylthiazol-2-yl)acetamide hydrobromide, 2-[(6-Methyl-2-pyridinyl)amino]-N-(5-methyl-2-thiazolyl)-4-thiazoleacetamide hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H15N5OS2HBr
Molecular Weight:
426.35
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

Biochem/physiol Actions

CDK12 kinase domain-targeting molecular glue that impairs CDK12/13 downstream signaling by inducing cyclin K (CycK) ubiquitination and degradation.
HQ461 is a CDK12 kinase domain-targeting molecular glue that promotes CDK12 interaction with DDB, resulting in DDB1-CUL4-RBX1 E3 ubiquitin ligase complex-mediated CDK12-interacting protein cyclin K (CycK) polyubiquitination and degradation (1-10 μM; A549). HQ461 treatment impairs A549 cellular CDK12 and CDK13 downstream signaling events, including RNA pol II C-terminal domain (CTD) Ser2 phosphorylation (1-10 μM) and DNA damage response genes expression (10 μM, 16 hrs).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service