Skip to Content
Merck
All Photos(1)

Key Documents

SML2728

Sigma-Aldrich

ML375

≥98% (HPLC)

Synonym(s):

(S)-9b-(4-Chlorophenyl)-1-(3,4-difluorobenzoyl)-1,2,3,9b-tetrahydro-5H-imidazo[2,1-a]isoindol-5-one, (S)-9b-(4-Chlorophenyl)-1-(3,4-difluorobenzoyl)-2,3-dihydro-1H-imidazo[2,1-a]isoindol-5(9bH)-one, ML 375, VU0483253

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H15ClF2N2O2
CAS Number:
Molecular Weight:
424.83
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -150 to -175, c = 0.5 in chloroform-d

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

Fc1c(ccc(c1)C(=O)N2[C@]4(N(CC2)C(=O)c5c4cccc5)c3ccc(cc3)Cl)F

InChI key

GXBAKXRLQAPKEE-QHCPKHFHSA-N

Biochem/physiol Actions

ML375 is an orally active, brain-penetrant, potent and M5 subtype-selective muscarinic acetylcholine receptor (mAChR) negative allosteric modulator (NAM) (IC50 = 300/790 nM against 10 μM NMS-induced Ca2+ mobilization in h/r M5-transfected CHO cells; h/r M1-M4 IC50 >30 μM) that reduces N-methylscopolamine (NMS) ML5 dissociation rate without competing against NMS for receptor binding. M375 shows in vivo efficacy in rat models of substances addiction (cocaine, ethanol, oxycodone) with excellent multispecies pharmacokinetic properties.
Orally active, brain-penetrant, potent & M5-selective muscarinic acetylcholine receptor (mAChR) NAM with in vivo efficacy in rat models of substances addiction.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alice E Berizzi et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 43(7), 1510-1517 (2018-02-28)
Despite the cost to both individual and society, alcohol use disorders (AUDs) remain a major health risk within society, and both relapse and heavy drinking are still poorly controlled with current medications. Here we demonstrate for the first time that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service