Skip to Content
Merck
All Photos(1)

Key Documents

SML1369

Sigma-Aldrich

Tesaglitazar

≥98% (HPLC)

Synonym(s):

(αS)-α-Ethoxy-4-[2-[4-[(methylsulfonyl)oxy]phenyl]ethoxy]-benzenepropanoic acid, (S)-2-Ethoxy-3-[4-[2-(4-methanesulfonyloxyphenyl)ethoxy]phenyl]propanoic acid, AR-H 039242, AZ 242, AZ-242, Tesaglitizar

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24O7S
CAS Number:
Molecular Weight:
408.47
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

O=S(OC1=CC=C(CCOC2=CC=C(C[C@H](OCC)C(O)=O)C=C2)C=C1)(C)=O

InChI

1S/C20H24O7S/c1-3-25-19(20(21)22)14-16-6-8-17(9-7-16)26-13-12-15-4-10-18(11-5-15)27-28(2,23)24/h4-11,19H,3,12-14H2,1-2H3,(H,21,22)/t19-/m0/s1

InChI key

CXGTZJYQWSUFET-IBGZPJMESA-N

Gene Information

Biochem/physiol Actions

Tesaglitazar is a potent and specific dual PPARα/γ agonist that improves insulin resistance and carbohydrate and lipid metabolism in ob/ob mice and obese Zucker rats.
Tesaglitazar is a potent and specific dual PPARα/γ agonist.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service