Skip to Content
Merck
All Photos(1)

Key Documents

SBR00089

Sigma-Aldrich

Sotorasib

≥98% (HPLC), solid

Synonym(s):

AMG-510

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H30F2N6O3
CAS Number:
Molecular Weight:
560.59
UNSPSC Code:
51101500

Quality Level

Assay

≥98% (HPLC)

form

solid

color

off-white to yellow

Mode of action

protein synthesis | inhibits

SMILES string

C[C@H]1CN(CCN1C2=NC(=O)N(C3=NC(=C(C=C32)F)C4=C(C=CC=C4F)O)C5=C(C=CN=C5C(C)C)C)C(=O)C=C

InChI

InChI=1S/C30H30F2N6O3/c1-6-23(40)36-12-13-37(18(5)15-36)28-19-14-21(32)26(24-20(31)8-7-9-22(24)39)34-29(19)38(30(41)35-28)27-17(4)10-11-33-25(27)16(2)3/h6-11,14,16,18,39H,1,12-13,15H2,2-5H3/t18-/m0/s1

InChI key

NXQKSXLFSAEQCZ-SFHVURJKSA-N

General description

Sotorasib (INN) is a small molecule which belongs to the class of compounds known as KRAS G12C inhibitors. Sotorasib is a chiral compound which is a KRAS G12C covalent inhibitor.

Biochem/physiol Actions

Sotorasib inhibits the KRAS G12C protein, which is involved in the regulation of cell growth and division. The KRAS G12C protein is a mutated form of the KRAS protein, which is found in certain types of cancer cells, particularly non-small cell lung cancer (NSCLC). The G12C mutation causes abnormal activation of the KRAS G12C protein, leading to the uncontrolled growth and spread of cancer cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service