Skip to Content
Merck
All Photos(1)

Key Documents

SBR00085

Sigma-Aldrich

Orbifloxacin

≥95% (H-NMR), powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20F3N3O3
CAS Number:
Molecular Weight:
395.38
UNSPSC Code:
51101500

Quality Level

Assay

≥95% (H-NMR)

form

powder

color

off-white to yellow

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

DNA synthesis | inhibits

storage temp.

2-30°C

SMILES string

C[C@@H]1CN(C[C@@H](N1)C)C2=C(C3=C(C(=C2F)F)C(=O)C(=CN3C4CC4)C(=O)O)F

InChI

InChI=1S/C19H20F3N3O3/c1-8-5-24(6-9(2)23-8)17-14(21)13(20)12-16(15(17)22)25(10-3-4-10)7-11(18(12)26)19(27)28/h7-10,23H,3-6H2,1-2H3,(H,27,28)/t8-,9+

InChI key

QIPQASLPWJVQMH-DTORHVGOSA-N

General description

Orbifloxacin is a fluoroquinolone antibiotic which inhibits bacterial growth by inhibiting DNA gyrase and topoisomerase IV.

Application

Orbifloxacin has very limited non-medical applications, as it is primarily known and used as a veterinary antibiotic for the treatment of bacterial infections in animals. However, it can be used as a reference standard in analytical chemistry for the development and validation of methods for the detection and quantification of fluoroquinolone antibiotics, such as Orbifloxacin, in various samples, such as food, environmental, and biological samples. Orbifloxacin has been used as a standard for the analysis of fluoroquinolone antibiotics in food products, such as meat, milk, and eggs, as well as in environmental samples, such as water and soil. Moreover, its potential use in animals, particularly livestock and poultry, highlights the importance of prudent use and management of the antibiotic to prevent the development and spread of antibiotic-resistant bacteria.

Biochem/physiol Actions

Orbifloxacin inhibits bacterial DNA synthesis through interference with the activity of DNA gyrase and topoisomerase IV enzymes, which are necessary for the replication, transcription, and repair of bacterial DNA. Orbifloxacin is active against Gram-negative and Gram-positive bacteria, including many strains of Escherichia coli, Pasteurella, Streptococcus, and Staphylococcus species.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service