Skip to Content
Merck
All Photos(1)

Key Documents

SAB5703046

Sigma-Aldrich

Anti-ICAM-1/CD54 Antibody, clone 20000000, Rabbit Monoclonal

Synonym(s):

BB2,CD54,P3.58,ICAM1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

2E7, monoclonal

form

liquid

species reactivity

human

concentration

1.00 mg/mL

technique(s)

immunofluorescence: 1:50 - 1:200

color

colorless

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ICAM1(3383)

General description

This gene encodes a cell surface glycoprotein which is typically expressed on endothelial cells and cells of the immune system. It binds to integrins of type CD11a / CD18, or CD11b / CD18 and is also exploited by Rhinovirus as a receptor. [provided by RefSeq, Jul 2008]

Immunogen

A synthesized peptide derived from human ICAM-1/CD54
PBS with 0.05% proclin300,0.05% BSA,50% glycerol,pH7.3.

Application

ELISA, FC,IF/ICC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service