SAB5702445
Anti-CD147/BSG Antibody, clone 5W2P7, Rabbit Monoclonal
Synonym(s):
5F7, CD147, EMMPRIN, OK, TCSF
Sign Into View Organizational & Contract Pricing
Select a Size
All Photos(1)
Select a Size
Change View
About This Item
UNSPSC Code:
12352203
NACRES:
NA.46
biological source
rabbit
Quality Level
material
colorless
clone
5W2P7, monoclonal
form
liquid
mol wt
40-60 kDa
species reactivity
mouse, mouse
concentration
0.5 mg/mL
technique(s)
western blot: 1:500 - 1:2000
color
colorless
General description
The protein encoded by this gene is a plasma membrane protein that is important in spermatogenesis, embryo implantation, neural network formation, and tumor progression. The encoded protein is also a member of the immunoglobulin superfamily. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Jul 2008]
Immunogen
A synthetic peptide corresponding to a sequence within amino acids 100-200 of human CD147/BSG (P35613).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.
Application
WB
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service