Skip to Content
Merck
All Photos(4)

Key Documents

SAB5702321

Sigma-Aldrich

Anti-LIS1 Antibody, clone 9O6X3, Rabbit Monoclonal

Synonym(s):

LIS1, LIS2, MDCR, MDS, NudF, PAFAH

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

9O6X3, monoclonal

form

liquid

mol wt

47 kDa

species reactivity

rat, rat, human

concentration

0.6 mg/mL

technique(s)

immunofluorescence: 1:50 - 1:200
immunoprecipitation (IP): 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

MVLSQRQRDELNRAIADYLRSNGYEEAYSVFKKEAELDVNEELDKKYAGLLEKKWTSVIRLQKKVMELESKLNEAKEEFTSGGPLGQKRDPKEWIPRPPE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PAFAH1B1(5048)

General description

This locus was identified as encoding a gene that when mutated or lost caused the lissencephaly associated with Miller-Dieker lissencephaly syndrome. This gene encodes the non-catalytic alpha subunit of the intracellular Ib isoform of platelet-activating factor acteylhydrolase, a heterotrimeric enzyme that specifically catalyzes the removal of the acetyl group at the SN-2 position of platelet-activating factor (identified as 1-O-alkyl-2-acetyl-sn-glyceryl-3-phosphorylcholine). Two other isoforms of intracellular platelet-activating factor acetylhydrolase exist: one composed of multiple subunits, the other, a single subunit. In addition, a single-subunit isoform of this enzyme is found in serum. [provided by RefSeq, Apr 2009]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 1-100 of human LIS1 (P43034).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB, IF/ICC, IP

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service