Skip to Content
Merck
All Photos(1)

Key Documents

SAB4702030

Monoclonal Anti-FOLR2 PE produced in mouse

new

Synonym(s):

BETA-HFR, FBP, FBP/PL-1, FR-BETA, FR-P3, Folate receptor beta

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

conjugate

phycoerythrin (R-PE) conjugate

Quality Level

antibody product type

primary antibodies

clone

EM-35, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

0.1 mg/ml

technique(s)

flow cytometry: suitable

isotype

IgG1

UniProt accession no.

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... FOLR2(2350)

General description

FOLR2 (folate receptor beta) is a cell surface protein that was originally thought to be specific for placenta, but it can be also expressed in other tissues, including hematopoietic cells. Its expression is increased in malignant tissues. FOLR2 may play a role in the transport of methotrexate in synovial macrophages in rheumatoid arthritis patients. FOLR2 is a marker for macrophages generated in the presence of M-CSF (M2), including M2-like tumor-associated macrophages, which exert potent immunosuppressive functions within the tumor environment, but not GM-CSF (M1), and whose expression correlates with increased folate uptake ability.

Specificity

The mouse monoclonal antibody EM-35 recognizes an extracellular epitope on FOLR2, a 30-40 kDa cell surface protein serving as a receptor for folic acid.

Immunogen

BW5147alpha,beta- cells

Physical form

Stabilizing phosphate buffered saline (PBS), pH 7.4, 15 mM sodium azide

Preparation Note

Purified antibody is conjugated with R-phycoerythrin (PE) under optimum conditions. Unconjugated antibody and free fluorochrome are removed by size-exclusion chromatography.

Storage and Stability

Store at 2-8°C. Protect from prolonged exposure to light. Do not freeze.

Other Notes

The antibody solution should be gently mixed before use.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service