Skip to Content
Merck
All Photos(1)

Key Documents

SAB4702026

Monoclonal Anti-CD279 PE produced in mouse

new

Synonym(s):

PD1, PDCD1, SLEB2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

conjugate

phycoerythrin (R-PE) conjugate

Quality Level

antibody product type

primary antibodies

clone

EH12.2H7, monoclonal

form

buffered aqueous solution

species reactivity

(Non-human primates), ( Human)

technique(s)

flow cytometry: suitable

isotype

IgG1κ

UniProt accession no.

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... PDCD1(5133)

General description

CD279 / PD-1 (programmed cell death 1), a transmembrane protein of CD28/CTLA-4 family. It is expressed inducibly mainly on activated T, B, and myeloid cells and plays a role in maintaining peripheral self-tolerance. Binding to its ligands CD273 and CD274 is associated with inhibition of T cell proliferation and induction of their anergy. It is also expressed during thymic development. Some variants of CD279 are associated with susceptibility to systemic lupus erythematosus, type 1 diabetes, and rheumatoid arthritis.

Specificity

The mouse monoclonal antibody EH12.2H7 recognizes an extracellular epitope of CD279 / PD-1 (programmed cell death 1), a 55 kDa type I transmembrane protein expressed above all during T cell development, on activated T cells, activated B cells, and activated monocytes.

Immunogen

recombinant human CD279

Physical form

Stabilizing phosphate buffered saline (PBS), pH 7.4, 15 mM sodium azide

Preparation Note

Purified antibody is conjugated with R-phycoerythrin (PE) under optimum conditions. Unconjugated antibody and free fluorochrome are removed by size-exclusion chromatography.

Storage and Stability

Store at 2-8°C. Protect from prolonged exposure to light. Do not freeze.

Other Notes

The antibody solution should be gently mixed before use.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service