SAB2108908
Anti-CRY1 (N-terminal) antibody produced in rabbit FITC conjugated
affinity isolated antibody
Synonym(s):
FRE, FRITZ, FRP-3, FRZB-1, FRZB-PEN
About This Item
biological source
rabbit
conjugate
FITC conjugate
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
64 kDa
species reactivity (predicted by homology)
mouse, zebrafish, canine, bovine, guinea pig, human, rat, horse, rabbit
concentration
0.5 mg/mL
technique(s)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 4-8 μg/mL
NCBI accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... CRY1(1407)
General description
Immunogen
Sequence
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service