Skip to Content
Merck
All Photos(2)

Key Documents

SAB1309051

[KO Validated]Anti-LAMP2 antibody produced in rabbit

new

[Knockout Validated],purified polyclonal antibody, buffered aqueous solution

Synonym(s):

Lysosome-associated membrane glycoprotein 2, LAMP-2, Lysosome-associated membrane protein 2, CD107 antigen-like family member B, CD107b, LAMP2, Knockout

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

(Human)

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

mol wt

52668Da

species reactivity

human

technique(s)

western blot: 1:8000

isotype

IgG

storage temp.

-10 to -25°C

target post-translational modification

unmodified

Gene Information

human ... LAMP2(3920)

General description

LAMP2 is a member of a family of membrane glycoproteins. This glycoprotein provides selectins with carbohydrate ligands. It may play a role in tumor cell metastasis. It may also function in the protection, maintenance, and adhesion of the lysosome.

Specificity

This LAMP2 antibody is generated from rabbits immunized with LAMP2 recombinant protein.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide. This antibody is purified through a protein A column, followed by peptide affinity purification.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service