SAB1309050
[KO Validated]Anti-Erk2 antibody produced in mouse

[Knockout Validated]purified monoclonal antibody, buffered aqueous solution
Synonym(s):
Mitogen-activated protein kinase 1, MAP kinase 1, MAPK 1, ERT1, Extracellular signal-regulated kinase 2, ERK-2, MAP kinase isoform p42, p42-MAPK, Mitogen-activated protein kinase 2, MAP kinase 2, MAPK 2, MAPK1, ERK2, PRKM1, PRKM2, Knockout
About This Item
biological source
(Human)
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
monoclonal
form
liquid
mol wt
83000Da
species reactivity
human, mouse
technique(s)
western blot: 1:1000
isotype
IgG1, kappa
storage temp.
-10 to -25°C
target post-translational modification
unmodified
Gene Information
human ... MAPK1(5594)
General description
Specificity
Physical form
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service