Skip to Content
Merck
All Photos(1)

Key Documents

SAB1309008

Anti-phospho-mouse CASP3(S26) antibody produced in rabbit

new

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Caspase-3, CASP-3, Apopain, Cysteine protease CPP32, CPP-32, LICE, Protein Yama, SREBP cleavage activity 1, SCA-1, Caspase-3 subunit p17, Caspase-3 subunit p12, Casp3, Cpp32

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

(Mouse)

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

mol wt

109343Da

species reactivity

mouse

technique(s)

dot blot: 1:500

isotype

IgG

NCBI accession no.

storage temp.

-10 to -25°C

target post-translational modification

phosphorylation (pSer26)

Gene Information

mouse ... Casp3(12367)

General description

This gene encodes a protein which is a member of thecysteine-aspartic acid protease (caspase) family. Sequentialactivation of caspases plays a central role in the execution-phaseof cell apoptosis. Caspases exist as inactive proenzymes whichundergo proteolytic processing at conserved aspartic residues toproduce two subunits, large and small, that dimerize to form theactive enzyme. This protein cleaves and activates caspases 6, 7 and9, and the protein itself is processed by caspases 8, 9 and 10. Itis the predominant caspase involved in the cleavage of amyloid-beta4A precursor protein, which is associated with neuronal death inAlzheimer′s disease. Alternative splicing of this gene results intwo transcript variants that encode the same protein. [provided byRefSeq].

Specificity

This mouse CASP3 Antibody is generated from rabbits immunized with a KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding S26 of mouse CASP3.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide. This antibody is purified through a protein A column, followed by peptide affinity purification.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service