Skip to Content
Merck
All Photos(1)

Key Documents

PZ0159

Sigma-Aldrich

MCOPPB trihydrochloride hydrate

>98% (HPLC)

Synonym(s):

1-[1-(1-Methylcyclooctyl)-4-piperidinyl]-2-[(3R)-3-piperidinyl]-1H-benzimidazole trihydrochloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H40N4 · 3 HCl · xH2O
CAS Number:
Molecular Weight:
518.01 (anhydrous basis)
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

>98% (HPLC)

form

powder

storage condition

desiccated

color

white to tan

solubility

H2O: ≥13 mg/mL

storage temp.

room temp

SMILES string

O.Cl.Cl.Cl.CC1(CCCCCCC1)N2CCC(CC2)n3c(nc4ccccc34)[C@@H]5CCCNC5

InChI

1S/C26H40N4.3ClH.H2O/c1-26(15-7-3-2-4-8-16-26)29-18-13-22(14-19-29)30-24-12-6-5-11-23(24)28-25(30)21-10-9-17-27-20-21;;;;/h5-6,11-12,21-22,27H,2-4,7-10,13-20H2,1H3;3*1H;1H2

InChI key

UQBVSGBDJOJHIP-UHFFFAOYSA-N

Biochem/physiol Actions

MCOPPB trihydrochloride is a potent non-peptide nociceptin/orphanin FQ peptide (NOP)-receptor full agonist [or opioid-receptor-like-1 (ORL1) receptor agonist]. MCOPPB is the most potent novel non-peptide NOP full agonist in vitro and an orally potent anxiolytic in the mice. It inhibited signaling through the NOP receptor in the mouse brain, suggesting that it penetrated into the brain after it was orally administered. It did not affect locomotor activity or memory, nor did it contribute to ethanol-induced hypnosis.
Potent non-peptide nociceptin/orphanin FQ peptide (NOP)-receptor full agonist [or opioid-receptor-like-1 (ORL1) receptor agonist.

Features and Benefits

This compound is featured on the Opioid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marco Raffaele et al.
GeroScience (2021-11-26)
Accumulation of senescent cells may drive age-associated alterations and pathologies. Senolytics are promising therapeutics that can preferentially eliminate senescent cells. Here, we performed a high-throughput automatized screening (HTS) of the commercial LOPAC®Pfizer library on aphidicolin-induced senescent human fibroblasts, to identify

Articles

We offers many products related to opioid receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service