Skip to Content
Merck
All Photos(2)

Key Documents

PRS4277

Sigma-Aldrich

Anti-ADAP (ab1) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Adhesion and degranulation adapter protein, Anti-FYB-120/130, Anti-FYN binding protein, Anti-FYN binding protein (FYB-120/130), Anti-SLAP-130

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse

technique(s)

immunocytochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FYB(2533)

Immunogen

a 20 amino acid peptide from near the carboxy-terminus of human ADAP.

Linkage

The action of this antibody can be blocked using blocking peptide SBP4277.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marcus Lettau et al.
Molecular immunology, 60(1), 72-79 (2014-04-29)
Nck adapter proteins link receptor and receptor-associated tyrosine kinases with proteins implicated in the regulation of the actin cytoskeleton. Nck is involved in a multitude of receptor-initiated signaling pathways and its physiological role thus covers aspects of tissue development and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service