Skip to Content
Merck
All Photos(1)

Key Documents

L6538

Sigma-Aldrich

Limaprost

≥99%, crystalline

Synonym(s):

11α,15S-Dihydroxy-17S,20-dimethyl-9-oxo-prosta-2E,13E-dien-1-oic acid, 17α,20-dimethyl-δ2-PGE1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H36O5
CAS Number:
Molecular Weight:
380.52
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥99%

form

crystalline

solubility

DMF: soluble
DMSO: soluble
ethanol: soluble

shipped in

wet ice

storage temp.

−20°C

SMILES string

CCCC[C@H](C)C[C@H](O)\C=C\[C@H]1[C@H](O)CC(=O)[C@@H]1CCCC\C=C\C(O)=O

InChI

1S/C22H36O5/c1-3-4-9-16(2)14-17(23)12-13-19-18(20(24)15-21(19)25)10-7-5-6-8-11-22(26)27/h8,11-13,16-19,21,23,25H,3-7,9-10,14-15H2,1-2H3,(H,26,27)/b11-8+,13-12+/t16-,17+,18+,19+,21+/m0/s1

InChI key

OJZYRQPMEIEQFC-UAWLTFRCSA-N

Related Categories

Biochem/physiol Actions

Limaprost exhibits cytoprotective, vasodilatory, and antiplatelet activity. Limaprost has the potential to treat lumbar spinal canal stenosis (LSCS) and several ischaemic symptoms of thromboangiitis obliterans (TAO).

Other Notes

Potent analog of PGE1 with prolonged half-life.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katsuhiko Nakai et al.
Prostaglandins, leukotrienes, and essential fatty acids, 69(4), 269-273 (2003-08-09)
An orally active prostaglandin E1 analogue, OP-1206 alpha-CD improves walking dysfunction in the rat spinal stenosis model. Loxoprofen-Na, a non-steroidal anti-inflammatory drug, is used to relieve chronic pain in patients with lumbar spinal canal stenosis. To determine whether the OP-1206
[A new oral prostaglandin E1 derivative (OP 1206) compared with intravenous PGE1: assessment of the effect on macro- and microcirculation].
P Scheffler et al.
VASA. Supplementum, 32, 386-391 (1991-01-01)
Katsuhiko Nakai et al.
Prostaglandins & other lipid mediators, 71(3-4), 253-263 (2003-10-02)
The systemic treatment effects of OP-1206 alpha-CD (17S-20-dimethyl-trans-delta 2-PGE1 alpha-cyclodextrin clathrate), a prostaglandin E1 (PGE1) analogue, on walking dysfunction, spinal cord blood flow (SCBF) and skin blood flow (SKBF) were assessed in the rat neuropathic intermittent claudication (IC) model in
P Ney et al.
Biomedica biochimica acta, 47(10-11), S186-S189 (1988-01-01)
The present study investigates the inhibitory action of the PGE1-analogue OP-1206 on stimulated LTB4 release from human PMN. In comparison with PGE2, PGE1 and iloprost, OP-1206 was equipotent to PGE2 and PGE1 as inhibitor of PAF (3 microM) and FMLP
T Kanayama et al.
The Journal of pharmacology and experimental therapeutics, 255(3), 1210-1217 (1990-12-01)
A chemically stable prostacyclin analog, KP-10614 [(4z,16s)-4,5,18,18,19,19-hexadehydro-16,20-dimethyl-delta 6(9;alpha)-9(o)- methano-PGI1], has been compared with two other prostacyclin derivatives (Iloprost and TEI-7165) and one prostaglandin E1 derivative (OP-1206) with respect to ADP-induced in vitro aggregation of human platelets and ex vivo platelet

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service