Skip to Content
Merck
All Photos(7)

Key Documents

HPA039325

Sigma-Aldrich

Anti-SDPR antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Cavin-2, Anti-Ps-p68, Anti-Sdr, Anti-Serum deprivation response

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:2500-1:5000

immunogen sequence

VSVERREKIKKSLTSNHQKISSGKSSPFKVSPLTFGRKKVREGESHAENETKSEDLPSSEQMPNDQEEESFAEGHSEASLASALVEGEIAEEAAEKATSRGSNSGMDSNIDLTIVEDEEEESVALEQAQKVRYE

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SDPR(8436)

General description

Serum deprivation protein response (SDPR) gene is mapped to human chromosome 2q32-q33. The gene codes for phosphatidylserine-binding protein, PS-p68.

Immunogen

serum deprivation response recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry. The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-SDPR antibody produced in rabbit has been used in the immunohistochemical and western blot analysis.

Biochem/physiol Actions

Serum deprivation protein response (SDPR) might be involved in platelet aggregation/secretion by facilitating the migration of platelet protein kinase C substrate, pleckstrin, to the plasma membrane in conjunction with phosphoinositides that bind to the C-terminal pleckstrin homology (PH) domain. SDPR assists binding of polymerase I and transcript release factor (PTRF) to caveolar membranes and plays a vital role in formation of caveolar invaginations. Thus, knock down or mutation of SDPR causes loss of caveolae and elevated expression of the protein leads to caveolae deformation and membrane tubulation. SDPR functions as a tumor suppressor in breast cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72701

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The platelet protein kinase C substrate pleckstrin binds directly to SDPR protein.
Baig A
Platelets, 20, 446-457 (2009)
The human serum deprivation response gene (SDPR) maps to 2q32-q33 and codes for a phosphatidylserine-binding protein.
Gustincich S
Genomics, 57, 120-129 (1999)
Increases in endothelial caveolin-1 and cavins correlate with cirrhosis progression.
Yokomori H
Micron (Oxford, England : 1993), 76, 52-61 (2015)
SDPR induces membrane curvature and functions in the formation of caveolae.
Hansen CG
Nature Cell Biology, 11, 807-814 (2009)
SDPR functions as a metastasis suppressor in breast cancer by promoting apoptosis.
Ozturk S
Proceedings of the National Academy of Sciences of the USA, 113, 638-643 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service