Skip to Content
Merck
All Photos(1)

Key Documents

EHU105771

Sigma-Aldrich

MISSION® esiRNA

targeting human HAX1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CACTTCGGGACTCAATGCTTAAGTATCCAGATAGTCACCAGCCCAGGATCTTTGGGGGGGTCTTGGAGAGTGATGCAAGAAGTGAATCCCCCCAACCAGCACCAGACTGGGGCTCCCAGAGGCCATTTCATAGGTTTGATGATGTATGGCCTATGGACCCCCATCCTAGAACCAGAGAGGACAATGATCTTGATTCCCAGGTTTCCCAGGAGGGTCTTGGCCCGGTTCTACAGCCCCAGCCCAAATCCTATTTCAAGAGCATCTCTGTGACCAAGATCACTAAACCAGATGGGATAGTGGAGGAGCGCCGGACTGTGGTGGACAGTGAGGGCCGGACAGAGACTACAGTAACCCGACACGAAGCAGATAGCAGTCCTAGGGGTGATCCAGAATCACCAAGACCTCCAGCCCTGGATGATGCCTTTTCCATCCTG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Li et al.
European review for medical and pharmacological sciences, 24(18), 9303-9314 (2020-10-06)
MicroRNA-325 (miR-325) is a tumor suppressor in some cancers. However, the role of miR-325 in determining the chemosensitivity to cancer cells is still not clear. The aim of this study was to investigate the effect of miR-325 on reversing the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service