Skip to Content
Merck
All Photos(1)

Key Documents

RENE 1

92092510, rat endometrium, Cuboidal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

RENE 1, 92092510

biological source

rat endometrium

packaging

tube of 5 μg 92092510-DNA-5UG
pkg of vial of cells 92092510-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Cuboidal

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat Sprague-Dawley, primary endometrial, adenovirus transformed

Cell Line Description

Derived from the endometrium tissue from the uterus of a 21-28 day old Sprague-Dawley rat. Cells carry vimentin, connexin 26 and connexin 43 markers. The transforming agent is adenovirus E1a protein.

Culture Medium

DMEM + 20μg/l Gly-His-Lys (Sigma) + 2mM Glutamine + 80 IU human Insulin/l (Novo) + 10% Foetal Bovine Serum (FBS) (heat-inactivated).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 i.e. seeding at 3x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C. Doubling time 48 hours.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service