Skip to Content
Merck
All Photos(1)

Key Documents

70Z/3

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

mouse blood

packaging

tube of 5 μg 91092002-DNA-5UG
pkg of vial of cells 91092002-1VL

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblastoid

products

Cytoplasmic Immunoglobulin

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse pre-B lymphocyte

Cell Line Description

Derived from a methyl nitrosourea-induced tumour in a (C57BL/6XDBA/2) F1 mouse. This line is positive for cytoplasmic Ig and negative for surface membrane Ig and TdT.

Culture Medium

RPMI 1640 + 2mM Glutamine + 0.05mM 2-Mercaptoethanol (2ME) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 2-5x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.
Depositor and originator: Dr P Kincade, Oklahoma Medical Research Foundation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service