Skip to Content
Merck
All Photos(1)

Key Documents

MC116

90110501, human lymph, Lymphoblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

MC116, 90110501

biological source

human lymph

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblast

products

Surface Immunoglobulin M (IgM), pentameric Immunoglobulin M (IgM)

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human lymphoma

Cell Line Description

Derived from the pleural effusion of a patient with undifferentiated lymphoma. The cells are EBNA negative, express surface IgM Lambda and secrete pentameric IgM which does not bind to protein A. Complement and Fc receptors were not detected.

Application

Hormone and tumourigenicity studies

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 10,12
D13S317: 12
D16S539: 12
D5S818: 12
D7S820: 10,13
THO1: 9.3,10
TPOX: 8,10
vWA: 15,17

Culture Medium

RPMI 1640 + 2mM Glutamine + 20% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 100,000 to 1,000,000 cells/ml; 5% CO2; 37°C. A maximum cell density of 2-4x1,000,000 cells/ml is achieved.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service