Skip to Content
Merck
All Photos(1)

Key Documents

KARPAS 45

6072602, human bone marrow, Spherical cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

KARPAS 45, 06072602

biological source

human bone marrow

packaging

tube of 5 μg 06072602-DNA-5UG
pkg of vial of cells 06072602-1VL

growth mode

Suspension

karyotype

Hypotetraploid

morphology

Spherical cells

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human T-cell Leukaemia

Cell Line Description

A human T cell leukaemia cell line established from the bone marrow of a 2-year-old boy diagnosed with acute lymphoblastic leukaemia (ALL) prior to therapy in 1972; cells were reported to carry the MLL-AFX and AFX-MLL fusion genes, the original diagnosis was T-ALL.

Culture Medium

RPMI 1640 + 2mM Glutamine + 20% Foetal Bovine Serum (FBS). For slower growth use 10% or 5% FBS.

Subculture Routine

Maintain cultures between 0.5 - 2 x1000,000 cells/ml; 5% CO2; 37°C. Doubling time 50-100 hours. Split saturated culture 1:2 to 1:3 every 2-3 days. Spherical cells growing in clusters in suspension.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service