Skip to Content
Merck
All Photos(8)

Key Documents

AMAB91818

Sigma-Aldrich

Anti-MLANA antibody produced in mouse

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, clone CL12874, purified by using Protein A, buffered aqueous glycerol solution

Synonym(s):

MART1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody product type

primary antibodies

clone

CL12874, monoclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

purified by

using Protein A

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 2-10 μg/mL
immunohistochemistry: 1:200-1:500
western blot: 1 μg/mL

isotype

IgG1

Ensembl | human accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MLANA(2315)

Immunogen

Melan-a

Application

All Prestige Antibodies® Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Features and Benefits

All Prestige Antibodies® Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Sequence

RRRNGYRALMDKSLHVGTQCALTRRCPQEGFDHRDSKVSLQEKNCEPVVPNAPPAYEKLSAEQSPPPYSP

Linkage

Corresponding Antigen APREST85192

Physical form

The antibodies are delivered in 40% glycerol and PBS (pH 7.2). 0.02% sodium azide is added as preservative.

Storage and Stability

Store at +4°C for short term storage. Long time storage is recommended at -20°C.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service