141-500
EpiVita Growth Medium for Neonatal and Fetal Keratinocytes, Serum-Free
Synonym(s):
Human Epidermal Keratinocytes (HEK) Serum-Free Growth Medium for neonatal cells (500 ml)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352207
NACRES:
NA.71
Recommended Products
form
liquid
Quality Level
shelf life
6 mo.
manufacturer/tradename
Cell Applications, Inc
storage condition
protect from light
shipped in
wet ice
storage temp.
2-8°C
General description
All-in-one, ready-to-use growth medium for cell culture of primary keratinocytes, neonatal and fetal
Quality
Each lot was tested for the ability to support plating, spreading and proliferation.
Preparation Note
contained in protocol
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service