PHR8617
Oxazepam Related Compound B

certified reference material, pharmaceutical secondary standard
Synonym(s):
3-(Acetyloxy)-7-chloro-1,3-dihydro-5-phenyl-2H-1,4-benzodiazepin-2-one, Oxazepam EP Impurity B; (3RS)-7-chloro-2-oxo-5-phenyl-2,3-dihydro-1H-1,4-benzodiazepin-3-yl acetate, 7-Chloro-1,3-dihydro-3-hydroxy-5-phenyl-2H-1,4-benzodiazepin-2-one acetate, 7-Chloro-2-oxo-5-phenyl-2,3-dihydro-1H-benzodiazepin-3-yl acetate, Oxazepam acetate
About This Item
grade
certified reference material
pharmaceutical secondary standard
Quality Level
Agency
(Not traceable to any primary pharmacopeia standard)
API family
oxazepam
CofA
current certificate can be downloaded
packaging
pkg of 50 mg
application(s)
pharmaceutical
storage temp.
-10 to -25°C
SMILES string
Clc1cc2c(cc1)NC(=O)C(N=C2c3ccccc3)OC(=O)C
InChI
1S/C17H13ClN2O3/c1-10(21)23-17-16(22)19-14-8-7-12(18)9-13(14)15(20-17)11-5-3-2-4-6-11/h2-9,17H,1H3,(H,19,22)
InChI key
FYRWUTOZBRWYCS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Analysis Note
Other Notes
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service