Skip to Content
Merck
All Photos(1)

Key Documents

NIST1595A

Tripalmitin

new

NIST®SRM®595A

Synonym(s):

1,2,3-Propanetriol tris(hexadecanoate), Glyceryl tripalmitate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)14COOCH2]2CHOCO(CH2)14CH3
CAS Number:
Molecular Weight:
807.32
Beilstein:
1811188
MDL number:
UNSPSC Code:
12352200

packaging

pkg of 2 g

mp

66-67 °C (lit.)

format

neat

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCC

InChI

1S/C51H98O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-49(52)55-46-48(57-51(54)45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)47-56-50(53)44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h48H,4-47H2,1-3H3

InChI key

PVNIQBQSYATKKL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A unit of SRM 1595a consists of one bottle containing approximately 2 g of high purity tripalmitin powder.SRM 1595A_cert SRM 1595A _SDS

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service