Skip to Content
Merck
All Photos(1)

Key Documents

PS1008-M

Sigma-Aldrich

PhosphoDetect Anti-FAK (pTyr⁸⁶¹) Rabbit pAb

liquid, Calbiochem®

Synonym(s):

Anti-FADK1, Phospho-Specific (Tyr⁸⁶¹), Anti-pp125FAK, Phospho-Specific (Tyr⁸⁶¹), Anti-Protein Tyrosine Kinase 2, Phospho-Specific (Tyr⁸⁶¹), Anti-PTK2, Phospho-Specific (Tyr⁸⁶¹), FAK 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

mouse, human, chicken

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr861)

Gene Information

human ... PTK2(5747)

General description

Rabbit polyclonal antibody adsorbed against non-phosphopeptide corresponding to the immunogen phosphorylation site and a generic phosphotyrosine-containing peptide, followed by immunoaffinity chromatography. Recognizes the ~125 kDa FAK protein phosphorylated at Tyr861.
Recognizes the ~125 kDa FAK protein phosphorylated at Tyr861 in primary chick embryo fibroblasts transfected with human FAK.
This PhosphoDetect Anti-FAK (pTyr⁸⁶¹) Rabbit pAb is validated for use in Immunoblotting, Immunohistochemistry for the detection of FAK (pTyr⁸⁶¹).

Immunogen

a synthetic phosphopeptide surrounding the Tyr⁸⁶¹ phosphorylation site of human FAK

Warning

Toxicity: Irritant (B)

Physical form

In Dulbecco′s PBS (without Mg2+ and Ca2+), 50 % glycerol, 1 mg/ml BSA, pH 7.3 (±0.1).

Analysis Note

Positive Control
Chicken embryo fibroblasts (CEF) expressing FAK, plated on fibronectin

Other Notes

Eliceiri, B.P., et al. 2002. J. Cell. Biol.157, 149.
Hauck, C.R., et al. 2002. J. Biol. Chem.277, 12487.
Lacalle, R.A., et al. 2002. J. Cell. Biol.157, 277.
Lim, Y., et al. 2002. J. Biol. Chem.277, 12735.
Melendez, J., et al. 2002. J. Biol. Chem.227, 45203.
Rigacci, S., et al. 2002. J. Biol. Chem.277, 41631.
Datta, A., et al. 2001. Mol. Cell. Biol.21, 7295.
Nakamura, K., et al. 2001. Oncogene20, 2626.
Rocic, P., et al. 2001. J. Biol. Chem.276, 21902.
Slack, J.K., et al. 2001. Oncogene20, 1152.
Sieg, D.J., et al. 2000. Nature Cell. Biol.2, 249.
Vial, D., et al. 2000. J. Biol. Chem.275, 28269.
Wennerberg, K., et al 2000. Mol. Cell. Biol.20, 5758.
The immunogen sequence is 100% conserved in rat and Xenopus FAK but cross-reactivity has not been tested. This antibody is also reported to work for immunohistochemistry. Antibody should be titrated for optimal use in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service