MACGLTVP1
MAS-100 CG Ex® Validation Protocol
Letter format
All Photos(1)
About This Item
UNSPSC Code:
41116500
NACRES:
JA.11
Recommended Products
Quality Level
compatibility
for use with MAS-100 CG Ex® system
General description
MAS-100 CG Ex® validation protocol is designed as a ready to use step by step guide to perform the validation activities for the compressed gas sampling pumps. The validation protocol in A4 format ensures regulatory compliance of the pumps and guarantees consistent performance.
Application
MAS-100 CG Ex® validation protocol is used for the validation of MAS-100 CG Ex® pumps which samples compressed gas for microbial contamination.
Legal Information
MAS-100 CG Ex is a registered trademark of MBV AG