Skip to Content
Merck
All Photos(2)

Key Documents

MABS1754

Sigma-Aldrich

Anti-p38/SAPK2 Antibody, clone 2F11

clone 2F11, from mouse

Synonym(s):

Mitogen-activated protein kinase 14, CRK1, MAP kinase 14, MAP kinase p38 alpha, MAPK 14, MAX-interacting protein 2, Mitogen-activated protein kinase p38 alpha, p38a, p38alpha, p38MAPK, SAPK2a, Stress-activated protein kinase 2a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

2F11, monoclonal

species reactivity

human, mouse, rat

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Mapk14(26416)

General description

Mitogen-activated protein kinase 14 (EC 2.7.11.24; UniProt P47811; also known as CRK1, MAP kinase 14, MAP kinase p38 alpha, MAPK 14, MAX-interacting protein 2, Mitogen-activated protein kinase p38 alpha, p38a, p38alpha, p38MAPK, SAPK2a, Stress-activated protein kinase 2a) is encoded by the Mapk14 (also known as Crk1, Csbp1, Csbp2, Mxi2) gene (Gene ID 26416) in murine species. p38 mitogen-activated protein kinases (MAPKs) constitute a group of stress-activated serine/threonine-specific kinases (SAPKs), including p38α (MAPK14/SAPK2A), p38β (MAPK11/SAPK2B), p38γ (ERK6/MAPK12/SAPK3), and p38δ (MAPK13/SAPK4), that mediate cellular signal transduction and play a vital role in numerous biological processes. p38α and p38β are ubiquitously expressed, while p38γ and p38δ are differentially expressed depending on tissue type. Sequence comparisons show approximately 60% identity within the p38 group. but only 40-45% to the other three MAP kinase family groups, extracellular signal-regulated kinases (ERKs), c-jun N-terminal or stress-activated protein kinases (JNK/SAPK), and ERK/big MAP kinase 1 (BMK1). All four p38 kinases contain a conserved Thr-Gly-Tyr (TGY) dual phosphorylation motif and are activated by dual kinases called MAP kinase kinases (MKKs).

Specificity

Clone 2F11 recognizes both p38alpha/SAPK2a and p38beta/SAPK2b.

Immunogen

Full-length murine p38/SAPK2a GST fusion protein.

Application

Research Category
Signaling
Research Sub Category
MAP Kinases
This Anti-p38 Antibody, clone 2F11 is validated for use in Western Blotting for the detection of SAPK2.
Western Blotting Analysis: A 1:500 dilution from a representative lot detected p38/SAPK2 in 10 µg of mouse (C2C12, NIH/3T3, RAW264), rat (L6, PC12), and human (HEK293, HeLa, HepG2, HUVEC, Jurkat) cell lysates, and in 10 µg of mouse brain homogenate.
Western Blotting Analysis: Representative lots detected p38 in human A549 epidermoid carcinoma cell and WI38-VA13 fibroblast lysates (Song, J.Y., et al. (2010). J. Biol. Chem. 285(12):9067-9076; Song, J.Y., et al. (2009). Cancer Lett. 283(2):135-142).
Western Blotting Analysis: A representative lot detected p38 in mouse embryonic stem cell lysate (Wu, X., et al. (2007). Dev. Dyn. 236(10):2767-2778).

Quality

Evaluated by Western Blotting in A431 cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected p38/SAPK2 in 10 µg of A431 cell lysate.

Target description

~38 kDa observed.

Linkage

Replaces: 05-454

Physical form

Format: Purified
Protein G purified.
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gwo-Jen Liaw et al.
Scientific reports, 9(1), 1885-1885 (2019-02-15)
To guard genome integrity, response mechanisms coordinately execute the G2/M checkpoint in responding to stress. p38 MAPK is activated to prolong the G2 phase for completion of damage repair. Tlk activity is required for DNA repair, chromosome segregation and G2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service