MABE995
Anti-NRF-1 Antibody, clone R157.1.3H3
clone R157.1.3H3, from mouse
Synonym(s):
Nuclear respiratory factor 1, NRF-1, Alpha palindromic-binding protein, Alpha-pal, NRF-1, Nuclear respiratory factor 1
About This Item
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
R157.1.3H3, monoclonal
species reactivity
human
technique(s)
ChIP: suitable (ChIP-seq)
western blot: suitable
isotype
IgG2aκ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... NRF1(4899)
General description
Immunogen
Application
Epigenetics & Nuclear Function
Nuclear Receptors
Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot detected NRF-1 chromatin DNA binding activity using K562 chromatin preparations (ENCODE Data Coordination Center (DCC) accession: ENCAB000BLM).
ChIP-seq Analysis: A representative lot detected NRF-1 DNA binding motifs by ChIP-seq analysis using K562 chromatin preparations (ENCODE Data Coordination Center (DCC) accession: ENCAB000BLM).
Quality
Western Blotting Analysis: 0.5 µg/mL of this antibody detected NRF-1 in 10 µg of Raji cell lysate.
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service