Skip to Content
Merck
All Photos(1)

Key Documents

MABE428

Sigma-Aldrich

Anti-WDR5 Antibody, clone 7B11

ascites fluid, clone 7B11, from mouse

Synonym(s):

WD repeat-containing protein 5, BMP2-induced 3-kb gene protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

7B11, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2b

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... WDR5(11091)

General description

WD repeat-containing protein 5 (WDR5) or alternatively BMP2-induced 3kb gene protein, is encoded by the human gene named WDR5/BIG3 and is part of a family of WD-40 repeat proteins that appear to facilitate the formation of multi-protein complexes much like a sticky tape will bind a variety of components together. The WD repeat proteins are involved in a wide array of cellular functions and processes including cell cycle progression, homeobox gene activation, apoptosis, gene regulation and histone methylation. One of WDR5 best studied activities is as part of a multi-protein complex that facilitates the methylation of histones, particularly histone H3, through its interaction with MLL a histone methyltransferase. WDR5 is also expressed in developing bone cells including osteoblasts, chondrocytes, and marrow stromal cells and works to accelerate osteoblast differentiation and positively promotes Wnt signaling in developing bone tissues. EMD-Millipore’s Anti-WDR5 mouse monoclonal antibody has been tested in western blot using HeLa cell lysates.

Immunogen

Purified recombinant fragment of human WDR5 expressed in E. Coli.

Application

Anti-WDR5 Antibody, clone 7B11 is a highly specific mouse monoclonal antibody & that targets WDR5 & has been tested in western blotting.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:500-2,000 dilution of this antibody detected WDR5 in HeLa cell lysate.

Optimal working dilutions must be determined by end user.

Target description

~40 kDa observed

Physical form

Mouse monoclonal IgG2b ascitic fluid containing up to 0.1% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
HeLa cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service