Skip to Content
Merck
All Photos(1)

Key Documents

MABC1745

Sigma-Aldrich

Anti-ErbB-3/HER3 Antibody, clone NG33

Synonym(s):

EC:2.7.10.1, Proto-oncogene-like protein c-ErbB-3, Receptor tyrosine-protein kinase erbB-3, Tyrosine kinase-type cell surface receptor HER3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

NG33, monoclonal

mol wt

calculated mol wt 148.1 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100

technique(s)

ELISA: suitable
flow cytometry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1κ

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

storage temp.

-10 to -25°C

Gene Information

human ... ERBB3(2065)

Specificity

Clone NG33 is a mouse monoclonal antibody that detects ErbB-3/HER3. It targets an epitope within the extracellular domain.

Immunogen

A recombinant fragment corresponding to the extracellular domain of human ErbB-3/HER3 combined with the Fc domain of human IgG1.

Application

Quality Control Testing

Evaluated by ELISA with recombinant Human ErRB-3/HER3 protein.

ELISA Analysis: Various dilutions of this antibody detected full-length recombinant Human ErbB-3/HER3 protein.

Tested Applications

Inhibition Assay: A representative lot of this antibody inhibited neuregulin (NRG)-induced migration and proliferation of cancer cells. (Gaborit, N., et al. (2015). Proc Natl Acad Sci USA. 112(3):839-44).

Flow Cytometry Analysis: A representative lot detected ErbB-3/HER3 in Flow Cytometry application (Gaborit, N., et al. (2015). Proc Natl Acad Sci USA. 112(3):839-44).

ELISA Analysis: A representative lot detected ErbB-3/HER3 in ELISA applications (Gaborit, N., et al. (2015). Proc Natl Acad Sci USA. 112(3):839-44).

Immunoprecipitation Analysis: A representative lot immunoprecipitated ErbB-3/HER3 in Immunoprecipitation application (Gaborit, N., et al. (2015). Proc Natl Acad Sci USA. 112(3):839-44).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Receptor tyrosine-protein kinase erbB-3 (UniProt: P21860; also known as EC:2.7.10.1, Proto-oncogene-like protein c-ErbB-3, Tyrosine kinase-type cell surface receptor HER3) is encoded by the ERBB3 (also known as HER3) gene (Gene ID: 2065) in human. ErbB3 is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-19), which is subsequently cleaved off to generate the mature form that contains an extracellular domain (aa 20-643), a transmembrane domain (aa 644-664), and a cytoplasmic domain (aa 665-1342). Its extracellular domain is formed of two ligand-binding regions (L1 and L2) and two cysteine-rich regions (CR1 and CR2). In the absence of a ligand, the extracellular domain stays in a locked conformation and is stabilized by interactions between CR1 and CR2 regions that prevent its dimerization. Its tyrosine kinase domain is localized to amino acids 709-966 and shares homology with EGFR and ErbB2 kinase domain. However, the tyrosine kinase activity is impaired due to modified residues and its activation requires the formation of heterodimers with other EGFR family receptors. Known ligands for ErbB3 include neuregulins (NRG) and NRG1 and NRG2 appear to be the most active. Downstream signaling pathways activated include MAPK and PI3 Kinase pathways. ErbB3 has been implicated in cell division and survival and its expression is essential for cell proliferation in several tumor models and transformation of normal tissues. It has also been implicated in acquired resistance to EGFR-tyrosine kinase inhibitors. Clone NG33 is shown to compete with NRG for binding to ErbB3/HER3 and inhibits NRG-induced migration and proliferation of cancer cells. It is reported to induce faster, and more extensive receptor degradation compared to NRG. (Ref.: Kiavue, N., et al. (2020). Oncogene. 39(3); 487-502; Gaborit, N., et al. (2015). Proc. Natl. Acad. Sci. USA. 112(3); 839-844).

Physical form

Purified mouse monoclonal antibody IgG1 in PBS without preservatives.

Reconstitution

1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service