Skip to Content
Merck
All Photos(3)

Key Documents

MABC1220

Sigma-Aldrich

Anti-RGS2 Antibody, clone 9D9.1

clone 9D9.1, from mouse

Synonym(s):

Regulator of G-protein signaling 2, Cell growth-inhibiting gene 31 protein, G0/G1 switch regulatory protein 8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9D9.1, monoclonal

species reactivity

rat, human

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... RGS2(5997)

General description

Regulator of G-protein signaling 2 (UniProt: P41220; also known as RGS2, Cell growth-inhibiting gene 31 protein, G0/G1 switch regulatory protein 8) is encoded by the RGS2 (also known as G0S8, GIG31) gene (Gene ID: 5997) in human. RGS2 is shown to regulate G protein-coupled receptor signaling cascades. It inhibits signal transduction by increasing the GTPase activity of G protein alpha subunits, thereby driving them into their inactive GDP-bound form. It also plays a role in the regulation of blood pressure in response to signaling via G protein-coupled receptors and GNAQ. RGS2 is reported to play a role in regulating the constriction and relaxation of vascular smooth muscle and also has a protective effect against myocardial hypertrophy and atrial arrhythmias. It inhibits beta-adrenergic receptor-induced cardiomyocyte hypertrophy. Overexpression of RGS2 reduces the activity of isoproterenol-induced ERK1/2 and prevents AKT activation. Four isoforms of RGS2 have been reported that are produced by alternative splicing. (Ref.: Nunn, C et al. (2010). Cell Signal 22(8); 1231-1239).

Specificity

Clone 9D9.1 specifically detects Regulator of G-protein signaling 2 (RGS2) in human and rat cells. It targets an epitope within 82 amino acids from the N-terminal region.

Immunogen

GST/His-tagged recombinant fragment corresponding to 82 amino acids from the N-terminal region of human Regulator of G-protein signaling 2 (RGS2).

Application

Anti-RGS2, clone 9D9.1, Cat. No. MABC1220, is a mosue monoclonal antibody that detects RGS2 in human and rat cells and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Research Category
Apoptosis & Cancer
Western Blotting Analysis: 1 µg/mL from a representative lot detected RGS2 in 10 µg of HL-60 cell lysate.

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected RGS2 in human cerebral cortex, human pancreas, human lymphoma, and rat testis tissues.

Quality

Evaluated by Western Blotting in Raji cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected RGS2 in 10 µg of Raji cell lysate.

Target description

~25 kDa observed; 24.38 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service