AP1170
Anti-PAFAH1B1 Mouse mAb (2C12)
liquid, clone 2C12, Calbiochem®
Synonym(s):
Anti-LIS1, Anti-PAFAHA, Anti-Platelet-Activating Factor Acetylhydrolase 1B α Subunit, Anti-Platelet-Activating Factor Acetylhydrolase 1B α Subunit, Anti-LIS1, Anti-PAFAHA
About This Item
biological source
mouse
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
2C12, monoclonal
form
liquid
does not contain
preservative
species reactivity
human
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
avoid repeated freeze/thaw cycles
isotype
IgG2a
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... PAFAH1B1(5048)
General description
Immunogen
Application

ELISA (see comments)
Immunoblotting (1-5 g/ml)
Warning
Physical form
Reconstitution
Other Notes
Legal Information
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service