Skip to Content
Merck
All Photos(1)

Key Documents

ABE430

Sigma-Aldrich

Anti-methyl-CARM1 (Arg551) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Histone-arginine methyltransferase CARM1, Coactivator-associated arginine methyltransferase 1, Protein arginine N-methyltransferase 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

monomethylation (Arg551)

Gene Information

human ... CARM1(10498)

General description

CARM1, also known as Histone-arginine methyltransferase CARM1, Coactivator-associated arginine methyltransferase 1, Protein arginine N-methyltransferase 4, and encoded by the gene name CARM1 and PRMT4, catalyzes the transfer of a methyl group from S-adenosyl-L-methionine to arginine residues. CARM1 methylates (mono- and asymmetric dimethylation) the the guanidino nitrogens of arginyl residues in several proteins involved in DNA packaging, transcription regulation, pre-mRNA splicing, and mRNA stability. Upon gene activation CARM1 together with histone acetyltransferases from EP300/P300 and p160 families, methylates histone H3 at ′Arg-17′ (H3R17me), forming mainly asymmetric dimethylarginine (H3R17me2a), leading to activate transcription via chromatin remodeling. During myogenic transcriptional activation, acts together with NCOA3/ACTR as a coactivator for MEF2C. During monocyte inflammatory stimulation, acts together with EP300/P300 as a coactivator for NF-kappa-B. Acts as coactivator for PPARG, promotes adipocyte differentiation and the accumulation of brown fat tissue. Plays a role in the regulation of pre-mRNA alternative splicing by methylation of splicing factors. Also methylates arginine residues in RNA-binding proteins PABPC1, ELAVL1 and ELAV4, which may affect their mRNA-stabilizing properties and the half-life of their target mRNAs. CARM1 is overexpressed in prostate adenocarcinomas and high-grade prostatic intraepithelial neoplasia.

Immunogen

Epitope: Methylated R551
Linear peptide corresponding to human CARM1 (Arg551) methylated at R551.

Application

Immunoprecipitation Analysis: A representative lot immunoprecipitated methyl-CARM1 (Arg551) in vitro methylation of a control buffer, 15 mg nuclear lysates, and 15 mg cytoplasmic lysates from CARM1/MEF cells by 1 mg CARM1 WT and CARM1 R551K (Kuhn, P., et al. (2011). Nucleic Acids Research. 39(7):2717-2726).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors
This Anti-methyl-CARM1 (Arg551) | ABE430 antibody is validated for use in WB, IP for the detection of methyl-CARM1 (Arg551) | ABE430.

Quality

Evaluated by Western Blotting in HEK293T CARM1 KO cells restored with WT CARM1, but not in HEK293T CARM1 KO cells restored with R551K mutant CARM1 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected methyl-CARM1 (Arg551) in 10 µg of HEK293T CARM1 KO cells restored with WT CARM1, but not in HEK293T CARM1 KO cells restored with R551K mutant CARM1 cell lysate.

Target description

~62 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing PBS with 0.05% sodium azide with 10% glycerol.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service