AB8125
Anti-phospho-Integrin β1 (Tyr783) Antibody
serum, Chemicon®
Sign Into View Organizational & Contract Pricing
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43
biological source
rabbit
Quality Level
antibody form
serum
antibody product type
primary antibodies
clone
polyclonal
species reactivity
human
manufacturer/tradename
Chemicon®
technique(s)
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
phosphorylation (pTyr783)
Gene Information
human ... ITGB1(3688)
Specificity
Integrin B1 phosphorylated on Tyr783
Immunogen
C-terminal Peptide
Application
Anti-phospho-Integrin β1 (Tyr783) Antibody detects level of phospho-Integrin β1 (Tyr783) & has been published & validated for use in WB.
Research Category
Cell Structure
Cell Structure
Research Sub Category
Integrins
Integrins
Quality
Routinely evaluated by immunoblot.
Target description
120kDa
Physical form
Raw Rabbit Serum with 0.1% Sodium Azide
Storage and Stability
1 year at 4°C from date of shipment
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service