Skip to Content
Merck
All Photos(1)

Key Documents

553009

Sigma-Aldrich

PhosphoDetect Anti-c-Raf (pTyr³⁴⁰, Tyr³⁴¹) Rabbit pAb

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

rat, human, mouse

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr340/pTyr341)

Gene Information

human ... RAF1(5894)

General description

Note: 1 T = 1 test. Sufficient for 10 Western miniblots.
Rabbit polyclonal antibody adsorbed against non-phosphopeptide corresponding to the immunogen phosphorylation site, followed by immunoaffinity chromatography. Recognizes the ~74 KDa c-Raf protein phosphorylated on Tyr340/341.
Recognizes c-Raf phosphorylated at Tyr340 and Tyr341 in HEK293 cells overexpressing c-Raf.
This PhosphoDetect Anti-c-Raf (pTyr³⁴⁰, Tyr³⁴¹) Rabbit pAb is validated for use in WB, ICC for the detection of c-Raf (pTyr³⁴⁰, Tyr³⁴¹).

Immunogen

Human
a synthetic phosphopeptide corresponding to amino acids surrounding the Tyr³⁴⁰ and Tyr³⁴¹, phosphorylation sites of human c-Raf (Raf-1)

Application

Immunoblotting (1:1000)

Immunocytochemistry (see comments)

Warning

Toxicity: Standard Handling (A)

Physical form

In phosphate buffer (without Mg2+ and Ca2+), 1 mg/ml BSA, 50% glycerol, pH 7.3 (+/- 0.1).

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
HEK293 cells over expressing c-Raf, phosphorylated in vitro by Src.

Other Notes

The immunogen sequence is 100% conserved in mouse and rat, but cross-reactivity has not been tested. This antibody has also been reported to work for immunocytochemistry. Variables associated with assay conditions will dictate the proper working dilution.
Yan, F., and Polk, D.B. 2001. Cancer Res.61, 963.
Yip-Schneider, M.T., et al. 2000. Biochem. J.351, 151.
Mason, C.S. et al. 1999. EMBO J.18, 2137.
Bonner, T.I. et al. 1985. Mol. Cell. Biol.5, 1400.
Moelling, K. et al. 1984. Nature312, 558.
Rapp, U.R. et al. 1983. Proc. Natl. Acad. Sci. USA80, 4218.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service