Skip to Content
Merck
All Photos(1)

Key Documents

5.05222

Sigma-Aldrich

RuBi-Nicotine

Synonym(s):

RuBi-Nicotine, Bis(2,2′-bipyridine-N,N′)bis(S-nicotine-N1) ruthenium(2+) dichloride complex Bis(2,2ʹ-bipyridine-N,Nʹ)bis(S-nicotine-N1) ruthenium(2+) dichloride complex , Nicotinic Cholinergic Receptor Agonist, RuBi-Nicotine, nAChR Receptor Agonist, RuBi-Nico

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H44Cl2N8Ru
CAS Number:
Molecular Weight:
808.81
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (NMR)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

deep orange

solubility

H2O: 100 mM

storage temp.

−20°C

SMILES string

[Ru+2]85([NH]9=CC=CC=C9C%10=CC=CC=[NH]%108)([NH]6=CC=CC=C6C7=CC=CC=[NH]75)([NH]3=CC=CC(=C3)[C@H]4N(CCC4)C)[NH]1=CC=CC(=C1)[C@H]2N(CCC2)C.[Cl-].[Cl-]

InChI

1S/2C10H14N2.2C10H8N2.2ClH.Ru/c2*1-12-7-3-5-10(12)9-4-2-6-11-8-9;2*1-3-7-11-9(5-1)10-6-2-4-8-12-10;;;/h2*2,4,6,8,10H,3,5,7H2,1H3;2*1-8H;2*1H;/q;;;;;;+2/p-2/t2*10-;;;;;/m00...../s1

InChI key

BQXVYFYPLMVART-ZPFSJBFKSA-L

General description

A highly water soluble Ruthenium-bisbipyridine-caged nicotine that can be un-caged by blue and green light. Rapid nicotine release (<20 ns) and fast biological effects in vivo.

Biochem/physiol Actions

Primary Target
nAChR

Warning

Toxicity: Toxic (F)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Filevich, O., et al. 2010. J. Inorg. Biochem.104, 51.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service